Assay Details
| Gene Symbol: dhc-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073871.3 | NP_506272.3 | 20 - 21 | 5242 | 163 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dhc-3| Gene Symbol: dhc-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_073871.3 | NP_506272.3 | 20 - 21 | 5242 | 163 | |
Set Membership: |
|