Assay Details
| Gene Symbol: sri-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269552.1 | NP_001256481.1 | 4 - 5 | 625 | 81 | |
| NR_069862.1 | 4 - 5 | 543 | 81 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sri-6| Gene Symbol: sri-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269552.1 | NP_001256481.1 | 4 - 5 | 625 | 81 | |
| NR_069862.1 | 4 - 5 | 543 | 81 | |||
Set Membership: |
|