Assay Details
| Gene Symbol: eea-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028956.4 | NP_001024127.1 | 4 - 5 | 1241 | 88 | |
| NM_001028957.5 | NP_001024128.1 | 4 - 5 | 1235 | 88 | ||
| GenBank mRNA | AY032860.1 | - | 4 - 5 | 1236 | 88 | |
More Information
Set Membership: |
|

eea-1
Genomic Map