Assay Details
Gene Symbol: cav-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269594.2 | NP_001256523.1 | 5 - 6 | 538 | 153 | |
NM_001269595.3 | NP_001256524.1 | 3 - 4 | 257 | 153 | ||
NM_001269596.3 | NP_001256525.1 | 4 - 5 | 371 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
