Assay Details
Gene Symbol: cav-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269594.2 | NP_001256523.1 | 7 - 8 | 745 | 69 | |
NM_001269595.3 | NP_001256524.1 | 5 - 6 | 464 | 69 | ||
NM_001269596.3 | NP_001256525.1 | 6 - 7 | 578 | 69 | ||
NM_001269597.3 | NP_001256526.1 | 1 - 2 | 101 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
