Assay Details
Gene Symbol: cav-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269594.2 | NP_001256523.1 | 8 - 9 | 812 | 120 | |
NM_001269595.3 | NP_001256524.1 | 6 - 7 | 531 | 120 | ||
NM_001269596.3 | NP_001256525.1 | 7 - 8 | 645 | 120 | ||
NM_001269597.3 | NP_001256526.1 | 2 - 3 | 168 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
