Assay Details
| Gene Symbol: ric-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047667.5 | NP_001041132.1 | 10 - 11 | 1755 | 83 | |
| NM_001047668.6 | NP_001041133.1 | 10 - 11 | 1649 | 83 | ||
More Information
Set Membership: |
|

ric-7
Genomic Map