Assay Details
| Gene Symbol: ric-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047667.5 | NP_001041132.1 | 4 - 5 | 659 | 88 | |
| NM_001047668.6 | NP_001041133.1 | 4 - 5 | 553 | 88 | ||
More Information
Set Membership: |
|

ric-7
Genomic Map