Assay Details
| Gene Symbol: ddx-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047669.6 | NP_001041134.1 | 4 - 5 | 1444 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ddx-17| Gene Symbol: ddx-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047669.6 | NP_001041134.1 | 4 - 5 | 1444 | 111 | |
Set Membership: |
|