Assay Details
| Gene Symbol: par-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028847.4 | NP_001024018.1 | 8 - 9 | 1605 | 63 | |
| NM_001047680.4 | NP_001041145.1 | 5 - 6 | 1215 | 63 | ||
| NM_001129495.4 | NP_001122967.2 | 5 - 6 | 1413 | 63 | ||
| NM_001269630.3 | NP_001256559.1 | 5 - 6 | 1338 | 63 | ||
| NM_001269632.3 | NP_001256561.1 | 4 - 5 | 1152 | 63 | ||
| NM_001269633.3 | NP_001256562.1 | 1 - 2 | 144 | 63 | ||
| NM_001307761.3 | NP_001294690.1 | 5 - 6 | 1614 | 63 | ||
| NM_001373155.1 | NP_001359744.1 | 5 - 6 | 1413 | 63 | ||
| NM_001380808.2 | NP_001368670.1 | 8 - 9 | 1902 | 63 | ||
More Information
Set Membership: |
|

par-1
Genomic Map