Assay Details
| Gene Symbol: ints-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269640.4 | NP_001256569.1 | 3 - 4 | 597 | 144 | |
| NM_001269641.2 | NP_001256570.1 | 3 - 4 | 602 | 144 | ||
More Information
Set Membership: |
|

ints-2
Genomic Map