Assay Details
| Gene Symbol: nxf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136408.3 | NP_001129880.1 | 7 - 8 | 1010 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nxf-1| Gene Symbol: nxf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136408.3 | NP_001129880.1 | 7 - 8 | 1010 | 71 | |
Set Membership: |
|