Assay Details
| Gene Symbol: rfc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269677.2 | NP_001256606.1 | 6 - 7 | 1479 | 108 | |
| NR_069988.1 | 5 - 6 | 1302 | 108 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rfc-1| Gene Symbol: rfc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269677.2 | NP_001256606.1 | 6 - 7 | 1479 | 108 | |
| NR_069988.1 | 5 - 6 | 1302 | 108 | |||
Set Membership: |
|