Assay Details
| Gene Symbol: rfc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269677.2 | NP_001256606.1 | 8 - 9 | 2186 | 65 | |
| NR_069988.1 | 7 - 8 | 2009 | 65 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rfc-1| Gene Symbol: rfc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269677.2 | NP_001256606.1 | 8 - 9 | 2186 | 65 | |
| NR_069988.1 | 7 - 8 | 2009 | 65 | |||
Set Membership: |
|