Assay Details
| Gene Symbol: rin-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129408.4 | NP_001122880.1 | 24 - 25 | 5580 | 82 | |
| NM_001129411.4 | NP_001122883.1 | 6 - 7 | 2605 | 82 | ||
| NM_001269690.3 | NP_001256619.1 | 23 - 24 | 5089 | 82 | ||
| NM_001269691.3 | NP_001256620.1 | 22 - 23 | 4774 | 82 | ||
| NM_001269692.3 | NP_001256621.1 | 16 - 17 | 3958 | 82 | ||
| NM_001269693.4 | NP_001256622.1 | 7 - 8 | 2760 | 82 | ||
| NM_001269694.3 | NP_001256623.1 | 4 - 5 | 1840 | 82 | ||
| NM_001276964.3 | NP_001263893.1 | 9 - 10 | 2914 | 82 | ||
| NM_001276965.3 | NP_001263894.1 | 7 - 8 | 2695 | 82 | ||
More Information
Set Membership: |
|

rin-1
Genomic Map