Assay Details
| Gene Symbol: rin-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129408.4 | NP_001122880.1 | 4 - 5 | 1033 | 105 | |
| NM_001269690.3 | NP_001256619.1 | 3 - 4 | 542 | 105 | ||
| NM_001269691.3 | NP_001256620.1 | 2 - 3 | 227 | 105 | ||
More Information
Set Membership: |
|

rin-1
Genomic Map