Assay Details
| Gene Symbol: ret-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129554.5 | NP_001123026.1 | 2 - 3 | 7924 | 156 | |
| NM_074255.7 | NP_506656.2 | 2 - 3 | 8025 | 156 | ||
More Information
Set Membership: |
|

ret-1
Genomic Map