Assay Details
| Gene Symbol: ret-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129554.5 | NP_001123026.1 | 5 - 6 | 8421 | 92 | |
| NM_001380820.1 | NP_001366847.1 | 3 - 4 | 499 | 92 | ||
| NM_074255.7 | NP_506656.2 | 5 - 6 | 8522 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380820.1 transcripts.
Set Membership: |
|

ret-1
Genomic Map