Assay Details
| Gene Symbol: ret-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029019.5 | NP_001024190.1 | 2 - 3 | 290 | 90 | |
| NM_001136438.3 | NP_001129910.1 | 3 - 4 | 1044 | 90 | ||
| NM_001380820.1 | NP_001366847.1 | 7 - 8 | 1740 | 90 | ||
| NM_001383456.1 | 7 - 8 | 1998 | 90 | |||
| NM_074255.7 | NP_506656.2 | 8 - 9 | 9560 | 90 | ||
| NM_074257.6 | NP_506658.1 | 2 - 3 | 416 | 90 | ||
More Information
Set Membership: |
|

ret-1
Genomic Map