Assay Details
| Gene Symbol: gcy-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074259.4 | NP_506660.2 | 15 - 16 | 3950 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

gcy-14| Gene Symbol: gcy-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074259.4 | NP_506660.2 | 15 - 16 | 3950 | 94 | |
Set Membership: |
|