Assay Details
| Gene Symbol: mut-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269709.2 | NP_001256638.1 | 8 - 9 | 1266 | 69 | |
| NM_001269710.3 | NP_001256639.1 | 5 - 6 | 889 | 69 | ||
More Information
Set Membership: |
|

mut-15
Genomic Map