Assay Details
| Gene Symbol: mut-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269709.2 | NP_001256638.1 | 9 - 10 | 1397 | 99 | |
| NM_001269710.3 | NP_001256639.1 | 6 - 7 | 1020 | 99 | ||
More Information
Set Membership: |
|

mut-15
Genomic Map