Assay Details
| Gene Symbol: sdc-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074302.3 | NP_506703.1 | 7 - 8 | 3956 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sdc-3| Gene Symbol: sdc-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074302.3 | NP_506703.1 | 7 - 8 | 3956 | 128 | |
Set Membership: |
|