Assay Details
| Gene Symbol: otub-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383461.1 | 2 - 3 | 1286 | 135 | ||
| Gene Symbol: C25D7.10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383462.1 | 3 - 4 | 1050 | 135 | ||
More Information
Set Membership: |
|

otub-1
Genomic Map