Assay Details
| Gene Symbol: sri-26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074358.2 | NP_506759.1 | 2 - 3 | 206 | 132 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sri-26| Gene Symbol: sri-26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074358.2 | NP_506759.1 | 2 - 3 | 206 | 132 | |
Set Membership: |
|