Assay Details
| Gene Symbol: cpr-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074389.5 | NP_506790.1 | 3 - 4 | 1104 | 60 | |
| GenBank mRNA | L39890.1 | - | 3 - 4 | 1080 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cpr-3| Gene Symbol: cpr-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074389.5 | NP_506790.1 | 3 - 4 | 1104 | 60 | |
| GenBank mRNA | L39890.1 | - | 3 - 4 | 1080 | 60 | |
Set Membership: |
|