Assay Details
| Gene Symbol: syp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074744.7 | NP_507145.2 | 4 - 5 | 2600 | 148 | |
| GenBank mRNA | AF515833.1 | - | 4 - 5 | 1111 | 148 | |
More Information
Set Membership: |
|

syp-1
Genomic Map