Assay Details
| Gene Symbol: srz-66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001356779.1 | NP_001343582.1 | 4 - 5 | 778 | 72 | |
| NR_002399.1 | 4 - 5 | 767 | 72 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

srz-66| Gene Symbol: srz-66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001356779.1 | NP_001343582.1 | 4 - 5 | 778 | 72 | |
| NR_002399.1 | 4 - 5 | 767 | 72 | |||
Set Membership: |
|