Assay Details
| Gene Symbol: skr-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074992.8 | NP_507393.1 | 1 - 2 | 77 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

skr-5| Gene Symbol: skr-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074992.8 | NP_507393.1 | 1 - 2 | 77 | 88 | |
Set Membership: |
|