Assay Details
| Gene Symbol: pgp-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075086.5 | NP_507487.1 | 5 - 6 | 1533 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pgp-9| Gene Symbol: pgp-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075086.5 | NP_507487.1 | 5 - 6 | 1533 | 82 | |
Set Membership: |
|