Assay Details
| Gene Symbol: ZK262.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075201.6 | NP_507602.1 | 6 - 7 | 1000 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZK262.2| Gene Symbol: ZK262.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075201.6 | NP_507602.1 | 6 - 7 | 1000 | 65 | |
Set Membership: |
|