Assay Details
| Gene Symbol: egas-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373187.2 | NP_001359624.1 | 11 | 1570 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

egas-1| Gene Symbol: egas-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373187.2 | NP_001359624.1 | 11 | 1570 | 85 | |
Set Membership: |
|