Assay Details
Gene Symbol: Y69H2.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129590.4 | NP_001123062.1 | 5 - 6 | 690 | 81 | |
NM_001276999.4 | NP_001263928.1 | 5 - 6 | 681 | 81 | ||
NM_001373189.3 | NP_001360619.1 | 5 - 6 | 698 | 81 | ||
NM_075243.8 | NP_507644.2 | 5 - 6 | 690 | 81 | ||
NR_023988.1 | 5 - 6 | 668 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
