Assay Details
| Gene Symbol: wars-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075282.6 | NP_507683.2 | 2 - 3 | 484 | 100 | |
| GenBank mRNA | AB096635.1 | - | 2 - 3 | 467 | 100 | |
More Information
Set Membership: |
|

wars-1
Genomic Map