Assay Details
| Gene Symbol: emb-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269902.2 | NP_001256831.1 | 2 - 3 | 407 | 94 | |
| NM_001313364.2 | NP_001300293.1 | 2 - 3 | 407 | 94 | ||
More Information
Set Membership: |
|

emb-4
Genomic Map