Assay Details
| Gene Symbol: ptr-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269905.3 | NP_001256834.1 | 5 - 6 | 1145 | 142 | |
| NM_001269906.3 | NP_001256835.1 | 2 - 3 | 514 | 142 | ||
More Information
Set Membership: |
|

ptr-22
Genomic Map