Assay Details
| Gene Symbol: ptr-22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269905.3 | NP_001256834.1 | 10 - 11 | 2531 | 72 | |
| NM_001269906.3 | NP_001256835.1 | 7 - 8 | 1900 | 72 | ||
More Information
Set Membership: |
|

ptr-22
Genomic Map