Assay Details
| Gene Symbol: F21D9.11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277007.2 | NP_001263936.1 | 2 - 3 | 167 | 124 | |
| NR_102122.1 | 2 - 3 | 327 | 124 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

F21D9.11| Gene Symbol: F21D9.11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277007.2 | NP_001263936.1 | 2 - 3 | 167 | 124 | |
| NR_102122.1 | 2 - 3 | 327 | 124 | |||
Set Membership: |
|