Assay Details
| Gene Symbol: abhd-3.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075462.7 | NP_507863.1 | 4 - 5 | 793 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

abhd-3.1| Gene Symbol: abhd-3.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075462.7 | NP_507863.1 | 4 - 5 | 793 | 101 | |
Set Membership: |
|