Assay Details
| Gene Symbol: sec-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075476.7 | NP_507877.1 | 2 - 3 | 237 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sec-23| Gene Symbol: sec-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075476.7 | NP_507877.1 | 2 - 3 | 237 | 96 | |
Set Membership: |
|