Assay Details
| Gene Symbol: fbxb-59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075498.4 | NP_507899.1 | 1 - 2 | 90 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fbxb-59| Gene Symbol: fbxb-59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075498.4 | NP_507899.1 | 1 - 2 | 90 | 69 | |
Set Membership: |
|