Assay Details
| Gene Symbol: mlt-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270009.3 | NP_001256938.1 | 14 - 15 | 8616 | 69 | |
| NM_001270010.3 | NP_001256939.1 | 13 - 14 | 7038 | 69 | ||
| NM_001270011.2 | NP_001256940.1 | 13 - 14 | 5777 | 69 | ||
| NM_001270012.3 | NP_001256941.1 | 14 - 15 | 8589 | 69 | ||
| NM_001270013.3 | NP_001256942.1 | 13 - 14 | 8254 | 69 | ||
| NM_001270014.3 | NP_001256943.1 | 14 - 15 | 6810 | 69 | ||
| NM_001270015.4 | NP_001256944.1 | 12 - 13 | 6404 | 69 | ||
| NM_001270016.3 | NP_001256945.1 | 12 - 13 | 6348 | 69 | ||
More Information
Set Membership: |
|

mlt-11
Genomic Map