Assay Details
| Gene Symbol: mlt-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270009.3 | NP_001256938.1 | 15 - 16 | 9236 | 78 | |
| NM_001270010.3 | NP_001256939.1 | 14 - 15 | 7658 | 78 | ||
| NM_001270011.2 | NP_001256940.1 | 14 - 15 | 6397 | 78 | ||
| NM_001270012.3 | NP_001256941.1 | 15 - 16 | 9209 | 78 | ||
| NM_001270013.3 | NP_001256942.1 | 14 - 15 | 8874 | 78 | ||
| NM_001270014.3 | NP_001256943.1 | 15 - 16 | 7430 | 78 | ||
| NM_001270015.4 | NP_001256944.1 | 13 - 14 | 7024 | 78 | ||
| NM_001270016.3 | NP_001256945.1 | 13 - 14 | 6968 | 78 | ||
More Information
Set Membership: |
|

mlt-11
Genomic Map