Assay Details
| Gene Symbol: mlt-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270009.3 | NP_001256938.1 | 4 - 5 | 693 | 92 | |
| NM_001270010.3 | NP_001256939.1 | 4 - 5 | 693 | 92 | ||
| NM_001270011.2 | NP_001256940.1 | 4 - 5 | 716 | 92 | ||
| NM_001270012.3 | NP_001256941.1 | 4 - 5 | 693 | 92 | ||
More Information
Set Membership: |
|

mlt-11
Genomic Map