Assay Details
| Gene Symbol: grl-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270026.2 | NP_001256955.1 | 1 - 2 | 384 | 93 | |
| NM_001270027.3 | NP_001256956.1 | 1 - 2 | 262 | 93 | ||
| NM_001270028.3 | NP_001256957.1 | 1 - 2 | 262 | 93 | ||
More Information
Set Membership: |
|

grl-14
Genomic Map