Assay Details
| Gene Symbol: mrps-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_075622.6 | NP_508023.1 | 2 - 3 | 215 | 100 | |
| Gene Symbol: exos-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383513.1 | 1 - 2 | 147 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383513.1 transcripts.
Set Membership: |
|

mrps-12
Genomic Map