Assay Details
| Gene Symbol: unc-80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028668.4 | NP_001023839.2 | 17 - 18 | 4086 | 103 | |
| NM_001028669.4 | NP_001023840.2 | 17 - 18 | 4339 | 103 | ||
| NM_001136423.4 | NP_001129895.2 | 17 - 18 | 4032 | 103 | ||
| NM_001270032.3 | NP_001256961.1 | 17 - 18 | 4086 | 103 | ||
More Information
Set Membership: |
|

unc-80
Genomic Map