Assay Details
| Gene Symbol: unc-80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028668.4 | NP_001023839.2 | 22 - 23 | 4818 | 88 | |
| NM_001136423.4 | NP_001129895.2 | 22 - 23 | 4764 | 88 | ||
| NM_001270032.3 | NP_001256961.1 | 23 - 24 | 4932 | 88 | ||
More Information
Set Membership: |
|

unc-80
Genomic Map