Assay Details
| Gene Symbol: unc-80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028668.4 | NP_001023839.2 | 35 - 36 | 9593 | 75 | |
| NM_001028669.4 | NP_001023840.2 | 33 - 34 | 9783 | 75 | ||
| NM_001136423.4 | NP_001129895.2 | 34 - 35 | 9503 | 75 | ||
| NM_001270032.3 | NP_001256961.1 | 36 - 37 | 9707 | 75 | ||
| NM_001270033.2 | NP_001256962.1 | 32 - 33 | 9398 | 75 | ||
More Information
Set Membership: |
|

unc-80
Genomic Map