Assay Details
| Gene Symbol: unc-80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028668.4 | NP_001023839.2 | 9 - 10 | 1912 | 97 | |
| NM_001028669.4 | NP_001023840.2 | 9 - 10 | 2165 | 97 | ||
| NM_001136423.4 | NP_001129895.2 | 9 - 10 | 1858 | 97 | ||
| NM_001270032.3 | NP_001256961.1 | 9 - 10 | 1912 | 97 | ||
| NM_001270033.2 | NP_001256962.1 | 9 - 10 | 1912 | 97 | ||
More Information
Set Membership: |
|

unc-80
Genomic Map